
Ketorolac trometamol

Synonym(s):
Ketorolac tris salt, (±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Toradol
Empirical Formula (Hill Notation):
C15H13NO3 · C4H11NO3
CAS Number:
Molecular Weight:
376.40
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing