
Nilotinib impurity B

Synonym(s):
Methyl 3-amino-4-methylbenzoate, Methyl 3-amino-p-toluate
Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing