Merck
  • Radical cation Diels-Alder cycloadditions by visible light photocatalysis.
Radical cation Diels-Alder cycloadditions by visible light photocatalysis.
Journal of the American Chemical Society (2011-10-29)
Shishi Lin, Michael A Ischay, Charles G Fry, Tehshik P Yoon
ABSTRACT
Ruthenium(II) polypyridyl complexes promote the efficient radical cation Diels-Alder cycloaddition of electron-rich dienophiles upon irradiation with visible light. These reactions enable facile [4 + 2] cycloadditions that would be electronically mismatched under thermal conditions. Key to the success of this methodology is the availability of ligand-modified ruthenium complexes that enable rational tuning of the electrochemical properties of the catalyst without significantly perturbing the overall photophysical properties of the system.
Sigma-Aldrich
1-Acetoxy-1,3-butadiene, mixture of cis and trans