Merck
  • Lipid hydroperoxide-induced and hemoglobin-enhanced oxidative damage to colon cancer cells.
Lipid hydroperoxide-induced and hemoglobin-enhanced oxidative damage to colon cancer cells.
Free radical biology & medicine (2011-05-24)
José Pedro F Angeli, Camila Carrião M Garcia, Fernanda Sena, Florencio P Freitas, Sayuri Miyamoto, Marisa Helena G Medeiros, Paolo Di Mascio
ABSTRACT
Epidemiological studies have indicated that Western diets are related to an increase in a series of malignancies. Among the compounds that are credited for this toxic effect are heme and lipid peroxides. We evaluated the effects of hemoglobin (Hb) and linoleic acid hydroperoxides (LAOOH) on a series of toxicological endpoints, such as cytotoxicity, redox status, lipid peroxidation, and DNA damage. We demonstrated that the preincubation of SW480 cells with Hb and its subsequent exposure to LAOOH (Hb + LAOOH) led to an increase in cell death, DCFH oxidation, malonaldehyde formation, and DNA fragmentation and that these effects were related to the peroxide group and the heme present in Hb. Furthermore, Hb and LAOOH alone exerted a toxic effect on the endpoints assayed only at concentrations higher than 100 μM. We were also able to show that SW480 cells presented a higher level of the modified DNA bases 8-oxo-7,8-dihydro-2'-deoxyguanosine and 1,N(2)-etheno-2'-deoxyguanosine compared to the control. Furthermore, incubations with Hb led to an increase in intracellular iron levels, and this high level of iron correlated with DNA oxidation, as measured as EndoIII- and Fpg-sensitive sites. Thus, Hb from either red meat or bowel bleeding could act as an enhancer of fatty acid hydroperoxide genotoxicity, which contributes to the accumulation of DNA lesions in colon cancer cells.
Sigma-Aldrich
2′-Deoxyguanosine monohydrate, 99-100%