Merck
  • Vascular endothelium-specific overexpression of human catalase in cloned pigs.
Vascular endothelium-specific overexpression of human catalase in cloned pigs.
Transgenic research (2010-12-21)
J J Whyte, M Samuel, E Mahan, J Padilla, G H Simmons, A A Arce-Esquivel, S B Bender, K M Whitworth, Y H Hao, C N Murphy, E M Walters, R S Prather, M H Laughlin
ABSTRACT
The objective of this study was to develop transgenic Yucatan minipigs that overexpress human catalase (hCat) in an endothelial-specific manner. Catalase metabolizes hydrogen peroxide (H(2)O(2)), an important regulator of vascular tone that contributes to diseases such as atherosclerosis and preeclampsia. A large animal model to study reduced endothelium-derived H(2)O(2) would therefore generate valuable translational data on vascular regulation in health and disease. Yucatan minipig fetal fibroblasts stably co-transfected with human catalase (Tie2-hCat) and eGFP expression constructs were isolated into single-cell populations. The presence of the Tie2-hCat transgene in individual colonies of fibroblasts was determined by PCR. Transgenic fibroblasts were used for nuclear transfer into enucleated oocytes by electrofusion. A minimum of 140 cloned embryos were transferred per surrogate sow (n = 4). All four surrogates maintained pregnancies and piglets were delivered by cesarean section. Nine male piglets from three of the four litters carried the Tie2-hCat transgene. Expression of human catalase mRNA and overall elevated catalase protein in isolated umbilical endothelial cells from transgenic piglets were verified by RT-PCR and western blot, respectively, and endothelial localization was confirmed by immunohistochemistry. Increased enzymatic activity of catalase in transgenic versus wild-type endothelial cells was inferred based on significantly reduced levels of H(2)O(2) in culture. The similarities in swine and human cardiovascular anatomy and physiology will make this pig model a valuable source of information on the putative role of endothelium-derived H(2)O(2) in vasodilation and in the mechanisms underlying vascular health and disease.
Sigma-Aldrich
Gentamicin sulfate salt, potency: ≥590 I.U. Gentamicin base per mg
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
15 mg
Estimated to ship on February 05, 2025
Details...
₩212,541