Merck
  • Effects of dietary nitrate supplementation on microvascular physiology at 4559 m altitude - A randomised controlled trial (Xtreme Alps).
Effects of dietary nitrate supplementation on microvascular physiology at 4559 m altitude - A randomised controlled trial (Xtreme Alps).
Nitric oxide : biology and chemistry (2019-10-12)
Andrew F Cumpstey, Philip J Hennis, Edward T Gilbert-Kawai, Bernadette O Fernandez, Daniel Grant, William Jenner, Matthieu Poudevigne, Helen Moyses, Denny Zh Levett, Alexandra Cobb, Paula Meale, Kay Mitchell, Helmut Pöhnl, Monty G Mythen, Michael Pw Grocott, Daniel S Martin, Martin Feelisch
ABSTRACT
Native highlanders (e.g. Sherpa) demonstrate remarkable hypoxic tolerance, possibly secondary to higher levels of circulating nitric oxide (NO) and increased microcirculatory blood flow. As part of the Xtreme Alps study (a randomised placebo-controlled trial of dietary nitrate supplementation under field conditions of hypobaric hypoxia), we investigated whether dietary supplementation with nitrate could improve NO availability and microvascular blood flow in lowlanders. Plasma measurements of nitrate, nitrite and nitroso species were performed together with measurements of sublingual (sidestream dark-field camera) and forearm blood flow (venous occlusion plethysmography) in 28 healthy adult volunteers resident at 4559 m for 1 week; half receiving a beetroot-based high-nitrate supplement and half receiving an identically-tasting low nitrate 'placebo'. Dietary supplementation increased plasma nitrate concentrations 4-fold compared to the placebo group, both at sea level (SL; 19.2 vs 76.9 μM) and at day 5 (D5) of high altitude (22.9 vs 84.3 μM, p < 0.001). Dietary nitrate supplementation also significantly increased both plasma nitrite (0.78 vs. 0.86 μM SL, 0.31 vs. 0.41 μM D5, p = 0.03) and total nitroso product (11.3 vs. 19.7 nM SL, 9.7 vs. 12.3 nM D5, p < 0.001) levels both at sea level and at 4559 m. However, plasma nitrite concentrations were more than 50% lower at 4559 m compared to sea level in both treatment groups. Despite these significant changes, dietary nitrate supplementation had no effect on any measured read-outs of sublingual or forearm blood flow, even when environmental hypoxia was experimentally reversed using supplemental oxygen. In conclusion, dietary nitrate supplementation does not improve microcirculatory function at 4559 m.
Sigma-Aldrich
Arginase Activity Assay Kit, sufficient for 200 colorimetric tests
Pricing and availability is not currently available.