Merck
  • Oncolytic Activity of Targeted Picornaviruses Formulated as Synthetic Infectious RNA.
Oncolytic Activity of Targeted Picornaviruses Formulated as Synthetic Infectious RNA.
Molecular therapy oncolytics (2020-06-13)
Noura B Elsedawy, Rebecca A Nace, Stephen J Russell, Autumn J Schulze
ABSTRACT
Infectious nucleic acid has been proposed as a superior formulation for oncolytic virus therapy. Oncolytic picornaviruses can be formulated as infectious RNA (iRNA), and their unwanted tropisms eliminated by microRNA (miRNA) detargeting. However, genomic insertion of miRNA target sequences into coxsackievirus A21 (CVA21) iRNA compromised its specific infectivity, negating further development as a novel oncolytic virus formulation. To address this limitation, we substituted a muscle-specific miRNA response element for the spacer region downstream of the internal ribosomal entry site in the 5' non-coding region of CVA21 iRNA, thereby preserving genome length while avoiding the disruption of known surrounding RNA structural elements. This new iRNA (R-CVA21) retained high specific infectivity, rapidly generating replicating miRNA-detargeted viruses following transfection in H1-HeLa cells. Further, in contrast with alternatively configured iRNAs that were tested in parallel, intratumoral administration of R-CVA21 generated a spreading oncolytic infection that was curative in treated animals without associated myotoxicity. Moreover, R-CVA21 also exhibited superior miRNA response element stability in vivo. This novel formulation is a promising agent for clinical translation.
Product Description
Roche
Titan One Tube RT-PCR System, sufficient for 100 reactions, kit of 1 (4 components), suitable for RT-PCR, hotstart: no