Merck
  • Quantifying the labeling and the levels of plant cell wall precursors using ion chromatography tandem mass spectrometry.
Quantifying the labeling and the levels of plant cell wall precursors using ion chromatography tandem mass spectrometry.
Plant physiology (2010-05-06)
Ana P Alonso, Rebecca J Piasecki, Yan Wang, Russell W LaClair, Yair Shachar-Hill
ABSTRACT
The biosynthesis of cell wall polymers involves enormous fluxes through central metabolism that are not fully delineated and whose regulation is poorly understood. We have established and validated a liquid chromatography tandem mass spectrometry method using multiple reaction monitoring mode to separate and quantify the levels of plant cell wall precursors. Target analytes were identified by their parent/daughter ions and retention times. The method allows the quantification of precursors at low picomole quantities with linear responses up to the nanomole quantity range. When applying the technique to Arabidopsis (Arabidopsis thaliana) T87 cell cultures, 16 hexose-phosphates (hexose-Ps) and nucleotide-sugars (NDP-sugars) involved in cell wall biosynthesis were separately quantified. Using hexose-P and NDP-sugar standards, we have shown that hot water extraction allows good recovery of the target metabolites (over 86%). This method is applicable to quantifying the levels of hexose-Ps and NDP-sugars in different plant tissues, such as Arabidopsis T87 cells in culture and fenugreek (Trigonella foenum-graecum) endosperm tissue, showing higher levels of galacto-mannan precursors in fenugreek endosperm. In Arabidopsis cells incubated with [U-(13)C(Fru)]sucrose, the method was used to track the labeling pattern in cell wall precursors. As the fragmentation of hexose-Ps and NDP-sugars results in high yields of [PO(3)](-)/or [H(2)PO(4)](-) ions, mass isotopomers can be quantified directly from the intensity of selected tandem mass spectrometry transitions. The ability to directly measure (13)C labeling in cell wall precursors makes possible metabolic flux analysis of cell wall biosynthesis based on dynamic labeling experiments.
Sigma-Aldrich
D-Galacto-D-mannan from Ceratonia siliqua, ~95% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
₩116,421
100 g
In Stock
Details...
₩233,450
250 g
In Stock
Details...
₩516,432
500 g
In Stock
Details...
₩963,641
1 kg
In Stock
Details...
₩1,612,125
5 kg
In Stock
Details...
₩5,589,402
25 kg
In Stock
Details...
₩11,934,066
50 kg
Estimated to ship on February 03, 2025
Details...
₩22,508,031