Merck
  • Degradation properties of a biodegradable shape memory elastomer, poly(glycerol dodecanoate), for soft tissue repair.
Degradation properties of a biodegradable shape memory elastomer, poly(glycerol dodecanoate), for soft tissue repair.
PloS one (2020-02-23)
Harsha Ramaraju, Loran D Solorio, Martin L Bocks, Scott J Hollister
ABSTRACT
Development of biodegradable shape memory elastomers (SMEs) is driven by the growing need for materials to address soft tissue pathology using a minimally invasive surgical approach. Composition, chain length and crosslinking of biocompatible polymers like PCL and PLA have been investigated to control mechanical properties, shape recovery and degradation rates. Depending on the primary mechanism of degradation, many of these polymers become considerably stiffer or softer resulting in mechanical properties that are inappropriate to support the regeneration of surrounding soft tissues. Additionally, concerns regarding degradation byproducts or residual organic solvents during synthesis accelerated interest in development of materials from bioavailable monomers. We previously developed a biodegradable SME, poly(glycerol dodecanoate) (PGD), using biologically relevant metabolites and controlled synthesis conditions to tune mechanical properties for soft tissue repair. In this study, we investigate the influence of crosslinking density on the mechanical and thermal properties of PGD during in vitro and in vivo degradation. Results suggest polymer degradation in vivo is predominantly driven by surface erosion, with no significant effects of initial crosslinking density on degradation time under the conditions investigated. Importantly, mechanical integrity is maintained during degradation. Additionally, shifts in melt transitions on thermograms indicate a potential shift in shape memory transition temperatures as the polymers degrade. These findings support the use of PGD for soft tissue repair and warrant further investigation towards tuning the molecular and macromolecular properties of the polymer to tailor degradation rates for specific clinical applications.
Sigma-Aldrich
Dodecanedioic acid, 99%