Merck
  • Oncogene-Induced Senescence Limits the Progression of Pancreatic Neoplasia through Production of Activin A.
Oncogene-Induced Senescence Limits the Progression of Pancreatic Neoplasia through Production of Activin A.
Cancer research (2020-06-20)
Yajie Zhao, Zhichong Wu, Marie Chanal, Fabienne Guillaumond, Delphine Goehrig, Sophie Bachy, Moitza Principe, Audrey Ziverec, Jean-Michel Flaman, Guillaume Collin, Richard Tomasini, Arja Pasternack, Olli Ritvos, Sophie Vasseur, David Bernard, Ana Hennino, Philippe Bertolino
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) is a deadly and aggressive cancer. Understanding mechanisms that drive preneoplastic pancreatic lesions is necessary to improve early diagnostic and therapeutic strategies. Mutations and inactivation of activin-like kinase (ALK4) have been demonstrated to favor PDAC onset. Surprisingly, little is known regarding the ligands that drive ALK4 signaling in pancreatic cancer or how this signaling pathway limits the initiation of neoplastic lesions. In this study, data mining and histologic analyses performed on human and mouse tumor tissues revealed that activin A is the major ALK4 ligand that drives PDAC initiation. Activin A, which is absent in normal acinar cells, was strongly induced during acinar-to-ductal metaplasia (ADM), which was promoted by pancreatitis or the activation of KrasG12D in mice. Activin A expression during ADM was associated with the cellular senescence program that is induced in precursor lesions. Blocking activin A signaling through the use of a soluble form of activin receptor IIB (sActRIIB-Fc) and ALK4 knockout in mice expressing KrasG12D resulted in reduced senescence associated with decreased expression of p21, reduced phosphorylation of H2A histone family member X (H2AX), and increased proliferation. Thus, this study indicates that activin A acts as a protective senescence-associated secretory phenotype factor produced by Kras-induced senescent cells during ADM, which limits the expansion and proliferation of pancreatic neoplastic lesions. SIGNIFICANCE: This study identifies activin A to be a beneficial, senescence-secreted factor induced in pancreatic preneoplastic lesions, which limits their proliferation and ultimately slows progression into pancreatic cancers.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
TGF-β RI Kinase Inhibitor VI, SB431542, InSolution, ≥97%