Merck
  • Improvement in Th1/Th2 immune homeostasis, antioxidative status and resistance to pathogenic E. coli on consumption of probiotic Lactobacillus rhamnosus fermented milk in aging mice.
Improvement in Th1/Th2 immune homeostasis, antioxidative status and resistance to pathogenic E. coli on consumption of probiotic Lactobacillus rhamnosus fermented milk in aging mice.
Age (Dordrecht, Netherlands) (2014-07-20)
Rohit Sharma, Rajeev Kapila, Gulshan Dass, Suman Kapila
ABSTRACT
Imbalance in Th1/Th2 immune pathways and cellular antioxidant systems with progressive aging are among the leading causes of increased risk of morbidity and mortality in elderly. Although probiotics have been considered to boost immune system, there is a lack of comprehensive analysis of probiotic effects on aging physiology. The present study aimed at determining anti-immunosenescence potential of milk fermented with probiotic Lactobacillus rhamnosus (LR) in 16 months old mice by concurrent analysis of immunosenescence markers associated with Th1/Th2 profile of splenocytes, inflamm-aging in plasma, neutrophil functions and antibody response in intestine along with analysis of antioxidant enzymes in liver and red blood cells (RBCs) after feeding trials of 1 and 2 months, respectively. An enteropathogenic Escherichia coli (ATCC 14948)-based infection model in aging mice was also designed to validate protective attributes of LR. Splenocytes registered increased IFN-γ and decreased IL-4 and IL-10 production in LR-fed animals. Neutrophil respiratory burst enzymes and phagocytosis increased significantly while no aggravation in plasma levels of MCP-1 and TNF-α was observed. Further, owing to increased Th1 response, antibodies registered a decrease in IgG1/IgG2a ratio and IgE levels in LR groups. No significant variations were observed in secretory IgA and IgA + cells in the intestine. Antioxidant enzymes (superoxide dismutase, catalase, glutathione peroxidase) in LR-fed groups recorded increased activities which were more pronounced in the liver than in RBCs. LR supplementation significantly reduced E. coli translocation to organs (intestine, liver, spleen, peritoneal fluid) by enhancing E. coli-specific antibodies (IgA and IgG1) and inflammatory proteins. In conclusion, LR supplementation alleviated immunosenescence-associated Th1/Th2 imbalance, improved antioxidant capacity, and enhanced resistance of aged mice to E. coli infection thereby signifying its potential in augmenting healthy aging.
Sigma-Aldrich
Cytochrome c Reductase (NADPH) Assay Kit, 1 kit sufficient for 100 tests, determining cytochrome c reductase activity