Merck
  • UV-LED as a New Emerging Tool for Curable Polyurethane Acrylate Hydrophobic Coating.
UV-LED as a New Emerging Tool for Curable Polyurethane Acrylate Hydrophobic Coating.
Polymers (2021-02-10)
Siti Khairunisah Ghazali, Nadia Adrus, Rohah A Majid, Fathilah Ali, Jamarosliza Jamaluddin
ABSTRACT
The elimination of mercury, low energy consumption, and low heat make the ultraviolet light-emitting diode (UV-LED) system emerge as a promising alternative to conventional UV-mercury radiation coating. Hence, a series of hydrophobic coatings based on urethane acrylate oligomer and fluorinated monomer via UV-LED photopolymerisation was designed in this paper. The presence of fluorine component at 1160 cm-1, 1235 cm-1, and 1296 cm-1 was confirmed by Fourier Transform Infra-Red spectroscopy. A considerably high degree C=C conversion (96-98%) and gel fraction (95-93%) verified the application of UV-LED as a new technique in radiation coating. It is well-accepted that fluorinated monomer can change the surface wettability as the water contact angle of the coating evolved from 88.4° to 121.2°, which, in turn, reduced its surface free energy by 70.5%. Hence, the hydrophobicity of the coating was governed by the migration of the fluorine component to the coating surface as validated by scanning electron and atomic force microscopies. However, above 4 phr of fluorinated monomer, the transparency of the cured coating examined by UV-visible spectroscopy experienced approximately a 16% reduction. In summary, the utilisation of UV-LED was a great initiative to develop green aspect in photopolymerisation, particularly in coating technology.
Sigma-Aldrich
Trimethylolpropane triacrylate, contains monomethyl ether hydroquinone as inhibitor, technical grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl methacrylate, contains MEHQ as inhibitor, 97%