Merck
  • [A newly discovered metabolic diseases due to defects in the pentose pathway].
[A newly discovered metabolic diseases due to defects in the pentose pathway].
Postepy biochemii (2011-09-15)
Anna Tylki-Szymańska, Teresa J Stradomska
ABSTRACT
Two previously unreported inborn errors of metabolism occur in the reversible part of the pentose phosphate pathway. Deficiency of ribose-5-phosphate isomerase has been described in one patient who suffered from a progressive leukoencephalopathy and developmental delay. Transaldolase deficiency has been diagnosed in 11 patients from 6 families in which the probands presented in the newborn and antenatal period with hepatospIenomegaly, hemolytic anaemia, hepatic fibrosis, kidney problems. Enzymes deficiency results in accumulations in body fluids erythritol, arabitol, ribitol, sedoheptitol, sedoheptulose, sedoheptulose-7-phosphate. Isomerase and transaldolase activity can be determined in leukocytes or fibroblasts.
Sigma-Aldrich
Phosphoriboisomerase from spinach, Type I, partially purified powder, ≥40 units/mg protein (biuret)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Estimated to ship on February 26, 2025
Details...
₩330,484
50 μg
Estimated to ship on February 26, 2025
Details...
₩589,817