Merck
  • Biodegradation of chloroacetamide herbicides by Paracoccus sp. FLY-8 in vitro.
Biodegradation of chloroacetamide herbicides by Paracoccus sp. FLY-8 in vitro.
Journal of agricultural and food chemistry (2011-03-23)
Jun Zhang, Jin-Wei Zheng, Bin Liang, Cheng-Hong Wang, Shu Cai, Ying-Ying Ni, Jian He, Shun-Peng Li
ABSTRACT
A butachlor-degrading strain, designated FLY-8, was isolated from rice field soil and was identified as Paracoccus sp. Strain FLY-8 could degrade and utilize six chloroacetamide herbicides as carbon sources for growth, and the degradation rates followed the order alachlor > acetochlor > propisochlor > butachlor > pretilachlor > metolachlor. The influence of molecular structure of the chloroacetamide herbicides on the microbial degradation rate was first analyzed; the results indicated that the substitutions of alkoxymethyl side chain with alkoxyethyl side chain greatly reduced the degradation efficiencies; the length of amide nitrogen's alkoxymethyl significantly affected the biodegradability of these herbicides: the longer the alkyl was, the slower the degradation efficiencies occurred. The phenyl alkyl substituents have no obvious influence on the degradation efficiency. The pathway of butachlor complete mineralization was elucidated on the basis of the results of metabolite identification and enzyme assays. Butachlor was degraded to alachlor by partial C-dealkylation and then converted to 2-chloro-N-(2,6-dimethylphenyl)acetamide by N-dealkylation, which subsequently transformed to 2,6-diethylaniline, which was further degraded via the metabolites aniline and catechol, and catechol was oxidized through an ortho-cleavage pathway. This study highlights an important potential use of strain FLY-8 for the in situ bioremediation of chloroacetamide herbicides and their metabolite-contaminated environment.
Sigma-Aldrich
2-Chloroacetamide, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Chloroacetamide, ≥98%