Merck
  • Discoloration of teeth after avulsion and replantation: results from a multicenter randomized controlled trial.
Discoloration of teeth after avulsion and replantation: results from a multicenter randomized controlled trial.
Journal of endodontics (2011-07-19)
Peter F Day, Monty S Duggal, Alec S High, Angus Robertson, Terry A Gregg, Paul F Ashley, Richard R Welbury, Ben O Cole, Stephen Westland
ABSTRACT
There is evidence to suggest that Ledermix, placed as an intervisit root canal dressing, might improve periodontal healing after replantation of avulsed teeth. As a part of a multicenter randomized controlled trial, we aimed to compare the effect of 2 root canal medicaments, Ledermix and Ultracal XS, on the discoloration of replanted teeth. Discoloration was investigated by using 3 methods: patient satisfaction with the color of replanted teeth, clinical photographs taken at baseline and 12-month reviews, and estimation of color change by using CIELAB scores for baseline and 12-month photographs. Twenty-two patients (27 teeth) were recruited. Ten patients (12 teeth) were randomized to the Ledermix group and 12 patients (15 teeth) to the Ultracal XS group. At 12 months, 8 patients were concerned with the discoloration of their teeth. Seven came from the Ledermix group and 1 from the Ultracal XS group. This difference was significant (Fisher exact test, P = .009). Standardized photographs were taken for the patients recruited at one center only (17 patients). There was significant discoloration of teeth from baseline with Ledermix, causing a darkening and gray-brown discoloration (mean change from baseline to 12 months, L∗ = -5.1, a∗ = 0.3, b∗ = -1.2, and ΔE = 8.1) and Ultracal XS, causing a yellowing and lightening of teeth (L∗ = 1.9, a∗ = 0.3, b∗ = 3.3, and ΔE = 5.4). There was a significant difference for the L∗ and b∗ variables (independent t test) between the 2 groups. Both root canal medicaments cause discoloration, with Ledermix proving less acceptable to patients.
Product Description
Supelco
Demeclocycline hydrochloride hydrate, VETRANAL®, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
₩255,577
5 g
In Stock
Details...
₩724,710
Sigma-Aldrich
Demeclocycline hydrochloride, powder or crystals
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
In Stock
Details...
₩255,577
5 g
In Stock
Details...
₩724,710