Merck
  • Natural lipid nanoparticles containing nimesulide: synthesis, characterization and in vivo antiedematogenic and antinociceptive activities.
Natural lipid nanoparticles containing nimesulide: synthesis, characterization and in vivo antiedematogenic and antinociceptive activities.
Journal of biomedical nanotechnology (2012-04-21)
Renata P Raffin, Amanda Lima, Ricardo Lorenzoni, Michelli B Antonow, Cláudia Turra, Marta P Alves, Solange B Fagan
ABSTRACT
Lipid nanoparticles are drug delivery systems able to increase bioavailability of poorly soluble drugs. They can be prepared with different lipid materials, especially natural lipids. Shea butter is a natural lipid obtained from the Butyrospermum parkii seed and rich in oleic and stearic acids. Nimesulide is a COX 2 selective anti-inflammatory that is poorly soluble in water. The purpose of this study was to develop and characterize shea butter lipid nanoparticles using a new technique and evaluate the in vivo activity of these nanoparticles. Lipid nanoparticles were prepared by melting shea butter and mixing with an aqueous phase using a high shear mixer. The nanoparticles presented pH of 6.9 +/- 0.1, mean particle size of 90 nm and a narrow polydispersity (0.21). Zeta potential was around -20 mV and the encapsulation efficiency was 97.5%. Drug release was evaluated using dialysis bags and presented monoexponential profile with t50% of 4.80 h (free drug t50% was only 2.86 h). Antinociceptive activity was performed by the acetic acid model. Both nimesulide and nimesulide-loaded nanoparticles presented significant activity compared to the control. The in vivo anti-inflammatory activity was evaluated by paw edema and was statistically different for the nanoparticles containing nimesulide compared to free nimesulide, blank nanoparticles and saline. In conclusion, the use of shea butter as encapsulating lipid was very successful and allowed nanoparticles to be prepared with a very simple technique. The nanoparticles presented significant pharmacological effects that were not seen for free drug administration.
Sigma-Aldrich
Nimesulide
Nimesulide, European Pharmacopoeia (EP) Reference Standard