Merck
  • A sensitive high performance liquid chromatographic assay of a new metabolite of ampicillin in man.
A sensitive high performance liquid chromatographic assay of a new metabolite of ampicillin in man.
The Journal of pharmacy and pharmacology (1986-03-01)
J Haginaka, J Wakai
ABSTRACT
A sensitive, high-performance liquid chromatographic method has been developed for the determination of piperazine-2,5-dione, a new metabolite of ampicillin, in human urine. Piperazine-2,5-dione was separated from human urine on a C18-column using phosphate buffer-methanol (pH 3.5) as eluent. Subsequently, the effluent underwent a postcolumn reaction with mercurous chloride and was then detected at 305 nm. It was found that piperazine-2,5-dione was also excreted in the urine of a volunteer dosed with ampicillin.
Sigma-Aldrich
Glycine anhydride, cyclic dipeptide
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
2.5 kU
Estimated to ship on January 27, 2025
Details...
₩92,264
10 kU
Estimated to ship on January 27, 2025
Details...
₩268,772