Merck
Characterization of 1,2-benzo-8-(2-aminoethyl)-3-phenoxazone.
Journal of pharmaceutical sciences (1987-08-01)
K G Bhansali, A M Kook
ABSTRACT
The compound 1,2-benzo-8-(2-aminoethyl)-3-phenoxazone (1) is synthesized by reacting tyramine with 1-nitroso-2-naphthol in the presence of nitric acid. During the reaction, tyramine and 1-nitroso-2-naphthol undergo condensation, ring closure, and subsequent oxidation steps to form 1. Compound 1 is characterized by one- and two-dimensional NMR spectroscopy and mass spectrometry. The proposed mechanism of reaction is based on the spectroscopic data. This reaction can be applied as a spot test analysis for analogous phenolic compounds.
Sigma-Aldrich
1-Nitroso-2-naphthol, 97%