MilliporeSigma
  • Expression of active human beta-glucuronidase in Sf9 cells infected with recombinant baculovirus.
Expression of active human beta-glucuronidase in Sf9 cells infected with recombinant baculovirus.
Life sciences (2002-07-20)
Bernhard Sperker, Thomas E Mürdter, Janne T Backman, Peter Fritz, Heyo K Kroemer
ABSTRACT
Antibody directed enzyme prodrug therapy (ADEPT) using glucuronide prodrugs is an experimental approach to reduce systemic toxicity of anti-cancer agents. Bioactivation of such prodrugs is achieved by fusion proteins consisting of targeting moieties (e.g. ligands of tumor specific antigens) and human beta-glucuronidase. In order to test a large panel of possible beta-glucuronidase fusion proteins for their applicability in ADEPT, an easy, rapid and high-yield expression system like the baculovirus/insect cell expression system would be needed. A prerequisite for using such fusion proteins is functional and biochemical characterization of human beta-glucuronidase expressed in baculovirus-infected insect cells. Therefore, recombinant human beta-glucuronidase was expressed in Sf9 insect cells and characterized at the protein and functional level. As shown by Western blot analysis the recombinant enzyme consists of dimers with their monomers being linked via disulfide bonds. Posttranslational modifications of the monomers seem to be different as compared with mammalian cells or tissues. The enzyme is functionally active in cleaving the substrates 5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, 4-methylumbelliferyl-beta-D-glucuronide and the glucuronide prodrug HMR 1826, respectively, with similar enzyme kinetic parameters as those found in human tissues. Our data demonstrate that beta-glucuronidase expressed in Sf9 cells displays the same enzymatic features as the protein expressed in mammalian cells. Therefore, we suggest that beta-glucuronidase fusion proteins produced in this cell line will be valuable tools for testing a large panel of various targeting moieties in human tumor xenograft models or may be used for ADEPT in man.
Product Description
Millipore
MUG Supplement, suitable for microbiology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 vials
Estimated to ship on March 29, 2025
$139.00