Merck
  • Recovery of Escherichia coli from soil after addition of sterile organic wastes.
Recovery of Escherichia coli from soil after addition of sterile organic wastes.
Applied and environmental microbiology (2006-03-07)
Adrian Unc, Julie Gardner, Susan Springthorpe
ABSTRACT
Laboratory batch tests indicate that addition of sterile municipal sewage biosolids to clay soil from four depths increases the numbers of Escherichia coli isolates recoverable in EC-MUG broth (EC broth with 4-methylumbelliferyl-beta-glucuronide). This effect was most marked for the deeper soil layers, with increases of about 2.6 orders of magnitude in E. coli most probable number.
Product Description
Millipore
MUG Supplement, suitable for microbiology
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 vials
Please contact Customer Service for Availability
₩221,575