Merck
  • Absence of the psbH gene product destabilizes the Photosystem II complex and prevents association of the Photosystem II-X protein in the thermophilic cyanobacterium Thermosynechococcus elongatus BP-1.
Absence of the psbH gene product destabilizes the Photosystem II complex and prevents association of the Photosystem II-X protein in the thermophilic cyanobacterium Thermosynechococcus elongatus BP-1.
Photosynthesis research (2006-05-16)
Masako Iwai, Mitsunori Katayama, Masahiko Ikeuchi
ABSTRACT
PS II-H is a small hydrophobic protein that is universally present in the PS II core complex of cyanobacteria and plants. The role of PS II-H was studied by directed mutagenesis and biochemical analysis in the thermophilic cyanobacterium Thermosynechococcus elongatus BP-1. The psbH disruptant could grow photoautotrophically; however, its growth was much slower than that of the wild type cell. Chromatography enabled the isolation of active oxygen-evolving PS II complexes from both the mutant and the wild type. The mutant yielded a relatively large amount of inactive PS II complex that lacked the following extrinsic proteins: the 33-kDa protein, the 12-kDa protein, and cytochrome c ( 550 ). There were differences between the psbH disruptant and the wild type in terms of the oxygen evolution activities of the cells, thylakoids, and PS II complexes. At high concentrations of 2,6-DCBQ, the activity was much lower in the mutant than in the wild type. Gel filtration chromatography of the PS II complexes showed that both active and inactive PS II complexes isolated from the mutant were mostly in the monomeric form, while the active PS II complex from the wild type was in the dimeric form. The polypeptide composition of both active and inactive PS II complexes from the mutant showed the absence of another small polypeptide, PS II-X. These results suggest that the PS II-H protein is essential for stable assembly of native dimeric PS II complex containing PS II-X.
Sigma-Aldrich
2,6-Dichloro-1,4-benzoquinone, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 units
In Stock
Details...
₩450,965
25 units
Please contact Customer Service for Availability
₩1,017,335
100 units
Please contact Customer Service for Availability
₩3,026,933