Merck
  • Rapid diagnosis of late-onset Pompe disease by fluorometric assay of alpha-glucosidase activities in dried blood spots.
Rapid diagnosis of late-onset Pompe disease by fluorometric assay of alpha-glucosidase activities in dried blood spots.
Molecular genetics and metabolism (2007-02-03)
Helmut Kallwass, Cortney Carr, Joseph Gerrein, Mariah Titlow, Robert Pomponio, Deeksha Bali, Jian Dai, Priya Kishnani, Alison Skrinar, Deyanira Corzo, Joan Keutzer
ABSTRACT
The enzymatic defect in Pompe disease is insufficient lysosomal acid alpha-glucosidase (GAA) activity which leads to lysosomal glycogen accumulation. We recently introduced a simple and reliable method to measure GAA activity in dried blood spots using Acarbose, a highly selective alpha-glucosidase inhibitor, to eliminate isoenzyme interference. Here we demonstrate that this method efficiently detects late-onset Pompe patients who are frequently misdiagnosed by conventional methods due to residual GAA activity in other tissue types.
Product Description
Millipore
MUG Supplement, suitable for microbiology