Merck
The metabolic functions and mechanism of action of vitamin K.
Scandinavian journal of clinical and laboratory investigation. Supplementum (1990-01-01)
L Uotila
ABSTRACT
Vitamin K functions in animal cells as the cofactor of the enzyme vitamin K-dependent carboxylase which catalyzes the post-translational formation of gamma-carboxyglutamyl (Gla) residues in specific vitamin K-dependent proteins. These proteins include four blood coagulation factors (prothrombin and Factors VII, IX and X), other plasma proteins (protein C, protein S and protein Z), two proteins from bone (osteocalcin or bone Gla-protein and matrix Gla-protein), and other proteins from lung, kidney, spleen, testis, placenta and other tissues. In the proteins involved in blood coagulation the Gla residues are mandatory for the activation of the inactive proenzymes; this process occurs on phospholipid surfaces to which the proenzymes are bound via Gla residues and calcium ions. The energy needed in the carboxylation reaction is obtained from the oxidation of vitamin K hydroquinone to 2,3-epoxide of the vitamin. Specific enzymes, vitamin K epoxide reductase and vitamin K quinone reductases, catalyze consecutive reactions in which the vitamin K hydroquinone is regenerated, thus allowing continued use of the vitamin K molecule for the carboxylations. The oral anticoagulants, derivatives of 4-hydroxycoumarin and indan-1,3-dione, used as therapeutic agents in thromboembolic disease, are antagonists to vitamin K preventing the catalytic use of vitamin K in the carboxylations by irreversibly inhibiting vitamin K epoxide reductase.
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Pricing and availability is not currently available.