Merck
  • Effect of telaprevir on the pharmacokinetics of buprenorphine in volunteers on stable buprenorphine/naloxone maintenance therapy.
Effect of telaprevir on the pharmacokinetics of buprenorphine in volunteers on stable buprenorphine/naloxone maintenance therapy.
Antimicrobial agents and chemotherapy (2012-05-09)
Xia Luo, Jose Trevejo, Rolf P G van Heeswijk, Frances Smith, Varun Garg
ABSTRACT
This was an open-label, single-sequence trial in hepatitis C virus-negative volunteers on stable, individualized, buprenorphine maintenance therapy. Telaprevir at 750 mg every 8 h was coadministered with buprenorphine/naloxone (4:1 ratio as sublingual tablets) for 7 days with food. Pharmacokinetic profiles of buprenorphine, norbuprenorphine, and naloxone were measured over the 24-hour dosing interval on day -1 (buprenorphine/naloxone alone, reference) and day 7 of telaprevir coadministration (test). Geometric least-squares mean ratios and associated 90% confidence intervals of treatment ratios (test/reference) were calculated using log-transformed pharmacokinetic parameters. Opioid withdrawal symptoms were evaluated throughout the study (via questionnaires and pupillometry). Pharmacokinetic data were available for 14 and 13 volunteers on day -1 and day 7, respectively. The area under the concentration-time curve (AUC) for buprenorphine was unchanged and the maximum concentration of drug in serum (C(max)) for buprenorphine, C(max) and AUC for norbuprenorphine, and C(max) naxolone were modestly decreased during coadministration with telaprevir. Geometric least-squares mean ratios (90% confidence intervals) for buprenorphine were 0.80 (0.69, 0.93) for the C(max) and 0.96 (0.84, 1.10) for the AUC from 0 to 24 h (AUC(0-24)); for norbuprenorphine, values were 0.85 (0.66, 1.09) for C(max) and 0.91 (0.71, 1.16) for AUC(0-24); for naloxone, the C(max) was 0.84 (0.62, 1.13). Coadministration of telaprevir did not increase withdrawal symptom frequency, and there were no serious adverse events reported during or after completion of telaprevir coadministration. Results suggest dose adjustment may not be necessary when telaprevir and buprenorphine/naloxone are coadministered.