Merck
  • Ganitumab with either exemestane or fulvestrant for postmenopausal women with advanced, hormone-receptor-positive breast cancer: a randomised, controlled, double-blind, phase 2 trial.
Ganitumab with either exemestane or fulvestrant for postmenopausal women with advanced, hormone-receptor-positive breast cancer: a randomised, controlled, double-blind, phase 2 trial.
The Lancet. Oncology (2013-02-19)
John Fr Robertson, Jean-Marc Ferrero, Hugues Bourgeois, Hagen Kennecke, Richard H de Boer, William Jacot, Jesse McGreivy, Samuel Suzuki, Min Zhu, Ian McCaffery, Elwyn Loh, Jennifer L Gansert, Peter A Kaufman
ABSTRACT
Insulin-like growth factors (IGF-1 and IGF-2) bind to the IGF-1 receptor (IGF-1R), increasing cell proliferation and survival. Ganitumab is a monoclonal IgG1 antibody that blocks IGF-1R. We tested the efficacy and safety of adding ganitumab to endocrine treatment for patients with hormone-receptor-positive breast cancer. We did this phase 2 trial in outpatient clinics and hospitals. We enrolled postmenopausal women with hormone-receptor-positive, locally advanced or metastatic breast cancer previously treated with endocrine treatment. They were randomly assigned (2:1) with a central randomisation schedule to receive intravenous ganitumab 12 mg per kg bodyweight or placebo in combination with open-label intramuscular fulvestrant (500 mg on day 1, then 250 mg on days 15, 29, and every 28 days) or oral exemestane (25 mg once daily) on a 28-day cycle. Patients, investigators, study monitors, and the sponsor staff were masked to treatment allocation. Response was assessed every 8 weeks. The primary endpoint was median progression-free survival in the intention-to-treat population. We analysed overall survival as one of our secondary endpoints. The study is registered at ClinicalTrials.gov, number NCT00626106. We screened 189 patients and enrolled 156 (106 in the ganitumab group and 50 in the placebo group). Median progression-free survival did not differ significantly between the ganitumab and placebo groups (3·9 months, 80% CI 3·6-5·3 vs 5·7 months, 4·4-7·4; hazard ratio [HR] 1·17, 80% CI 0·91-1·50; p=0·44). However, overall survival was worse in the the ganitumab group than in the placebo group (HR 1·78, 80% CI 1·27-2·50; p=0·025). With the exception of hyperglycaemia, adverse events were generally similar between groups. The most common grade 3 or higher adverse event was neutropenia-reported by six of 106 (6%) patients in the ganitumab group and one of 49 (2%) in the placebo group. Hyperglycaemia was reported by 12 of 106 (11%) patients in the ganitumab group (with six patients having grade 3 or 4 hyperglycaemia) and none of 49 in the placebo group. Serious adverse events were reported by 27 of 106 (25%) patients in the ganitumab group and nine of 49 (18%) patients in the placebo group. Addition of ganitumab to endocrine treatment in women with previously treated hormone-receptor-positive locally advanced or metastatic breast cancer did not improve outcomes. Our results do not support further study of ganitumab in this subgroup of patients. Amgen.
Sigma-Aldrich
Exemestane, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mg
Estimated to ship on January 20, 2025
Details...
₩193,053
10 mg
Estimated to ship on January 20, 2025
Details...
₩989,930