- Preparation and antigenic properties of estriol 16-glucuronide- and estradiol 17-glucuronide-[C-6]-bovine serum albumin conjugates.
Preparation and antigenic properties of estriol 16-glucuronide- and estradiol 17-glucuronide-[C-6]-bovine serum albumin conjugates.
Journal of steroid biochemistry (1982-04-01)
T Nambara, K Shimada, T Ohkubo, T Niwa
PMID7087478
ABSTRACT
The preparation and antigenic properties of estriol 16-glucuronide-bovine serum albumin (BSA) and estradiol 17-glucuronide-BSA conjugates in which the hapten molecule is linked to the carrier protein through an (O-carboxymethyl)oxime bridge at the C-6 position on the steroid nucleus, have been described. Antibodies raised against the two immunogens in the rabbit possessed excellent specificity to estriol 16-glucuronide and estradiol 17-glucuronide, respectively, exhibiting no significant cross-reactions with estrogen glucuronides having a substituent in ring A and no cross-reactivities with free estrogens, their sulfates and related steroids.