Merck
  • Molecular mechanism for metal-independent production of hydroxyl radicals by hydrogen peroxide and halogenated quinones.
Molecular mechanism for metal-independent production of hydroxyl radicals by hydrogen peroxide and halogenated quinones.
Proceedings of the National Academy of Sciences of the United States of America (2007-10-31)
Ben-Zhan Zhu, Balaraman Kalyanaraman, Gui-Bin Jiang
ABSTRACT
We have shown previously that hydroxyl radicals (HO*) can be produced by H2O2 and halogenated quinones, independent of transition metal ions; however, the underlying molecular mechanism is still unclear. In the present study, using the electron spin resonance secondary radical spin-trapping method, we found that tetrachloro-1,4-benzoquinone (TCBQ), but not its corresponding semiquinone anion radical, the tetrachlorosemiquinone anion radical (TCSQ*-), is essential for HO* production. The major reaction product between TCBQ and H2O2 was identified by electrospray ionization quadrupole time-of-flight mass spectrometry to be the ionic form of trichlorohydroxy-1,4-benzoquinone (TrCBQ-OH), and H2O2 was found to be the source and origin of the oxygen atom inserted into the reaction product TrCBQ-OH. On the basis of these data, we propose that HO* production by H2O2 and TCBQ is not through a semiquinone-dependent organic Fenton reaction but rather through the following mechanism: a nucleophilic attack of H2O2 to TCBQ, forming a trichlorohydroperoxyl-1,4-benzoquinone (TrCBQ-OOH) intermediate, which decomposes homolytically to produce HO*. This represents a mechanism of HO* production that does not require redox-active transition metal ions.
Sigma-Aldrich
Tetrafluoro-1,4-benzoquinone, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
110 g
Estimated to ship on March 24, 2025
$115.00