Merck
  • Mercaptopyridine-N-oxide, an NADH-fumarate reductase inhibitor, blocks Trypanosoma cruzi growth in culture and in infected myoblasts.
Mercaptopyridine-N-oxide, an NADH-fumarate reductase inhibitor, blocks Trypanosoma cruzi growth in culture and in infected myoblasts.
FEMS microbiology letters (1999-07-01)
J F Turrens, C L Newton, L Zhong, F R Hernandez, J Whitfield, R Docampo
ABSTRACT
The enzyme NADH-fumarate reductase is not found in mammalian cells but it is present in several parasitic protozoa including Trypanosoma cruzi, the parasite that causes Chagas' disease. This study shows that the drug 2-mercaptopyridine-N-oxide (MPNO) inhibits NADH-fumarate reductase purified from T. cruzi (ID50 = 35 microM). When added to intact cells, MPNO inhibited the growth of T. cruzi epimastigotes in culture (ID50 = 0.08 microM) as well as the infection of mammalian myoblasts by T. cruzi trypomastigotes (ID50 = 20 microM). At a concentration of 2.4 microM, MPNO also inhibited the growth of amastigotes (intracellular dividing forms) in cultured mammalian myoblasts. Supplementation of culture media with 5 mM succinate, the product of fumarate reductase, partially protected against the inhibition of the growth of epimastigotes by MPNO. Moreover, MPNO inhibited the accumulation of succinate in cultures of epimastigotes, as measured by high performance liquid chromatography. Although MPNO may have other intracellular targets in addition to fumarate reductase, these results support the hypothesis that compounds which inhibit the enzyme fumarate reductase may be potential chemotherapeutic agents against Chagas' disease.
Sigma-Aldrich
2-Mercaptopyridine N-oxide, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Please contact Customer Service for Availability
₩96,527
100 g
Please contact Customer Service for Availability
₩299,831
250 g
Please contact Customer Service for Availability
₩703,192
1 kg
Please contact Customer Service for Availability
₩2,134,850