- Fabrication and luminescence of monolayered boron nitride quantum dots.
Fabrication and luminescence of monolayered boron nitride quantum dots.
Small (Weinheim an der Bergstrasse, Germany) (2013-07-11)
Liangxu Lin, Yaoxian Xu, Shaowei Zhang, Ian M Ross, Albert C M Ong, Dan A Allwood
PMID23839969
ABSTRACT
Monolayered boron nitride (BN) quantum dots (QDs; lateral size ≈10 nm) are fabricated using a novel method. Unlike monolayered BN sheets, these BN QDs exhibit blue-green luminescence due to defects formed during preparation. This optical behavior adds significant functionality to a material that is already receiving much attention. It is further shown that the QDs are nontoxic to biological cells and well suited to bio-imaging.
MATERIALS