Merck
  • Phospholipase C-related catalytically inactive protein (PRIP) controls KIF5B-mediated insulin secretion.
Phospholipase C-related catalytically inactive protein (PRIP) controls KIF5B-mediated insulin secretion.
Biology open (2014-05-09)
Satoshi Asano, Tomomi Nemoto, Tomoya Kitayama, Kae Harada, Jun Zhang, Kana Harada, Isei Tanida, Masato Hirata, Takashi Kanematsu
ABSTRACT
We previously reported that phospholipase C-related catalytically inactive protein (PRIP)-knockout mice exhibited hyperinsulinemia. Here, we investigated the role of PRIP in insulin granule exocytosis using Prip-knockdown mouse insulinoma (MIN6) cells. Insulin release from Prip-knockdown MIN6 cells was higher than that from control cells, and Prip knockdown facilitated movement of GFP-phogrin-labeled insulin secretory vesicles. Double-immunofluorescent staining and density step-gradient analyses showed that the KIF5B motor protein co-localized with insulin vesicles in Prip-knockdown MIN6 cells. Knockdown of GABAA-receptor-associated protein (GABARAP), a microtubule-associated PRIP-binding partner, by Gabarap silencing in MIN6 cells reduced the co-localization of insulin vesicles with KIF5B and the movement of vesicles, resulting in decreased insulin secretion. However, the co-localization of KIF5B with microtubules was not altered in Prip- and Gabarap-knockdown cells. The presence of unbound GABARAP, freed either by an interference peptide or by Prip silencing, in MIN6 cells enhanced the co-localization of insulin vesicles with microtubules and promoted vesicle mobility. Taken together, these data demonstrate that PRIP and GABARAP function in a complex to regulate KIF5B-mediated insulin secretion, providing new insights into insulin exocytic mechanisms.
Sigma-Aldrich
Anti-KIF5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution