Merck
CypA, a gene downstream of HIF-1α, promotes the development of PDAC.
PloS one (2014-03-26)
Huan Zhang, Jing Chen, Fenghua Liu, Chuntao Gao, Xiuchao Wang, Tiansuo Zhao, Jingcheng Liu, Song Gao, Xiao Zhao, He Ren, Jihui Hao
ABSTRACT
Hypoxia-inducible factor-1α (HIF-1α) is a highly important transcription factor involved in cell metabolism. HIF-1α promotes glycolysis and inhibits of mitochondrial respiration in pancreatic ductal adenocarcinoma (PDAC). In response to tumor hypoxia, cyclophilin A (CypA) is over-expressed in various cancer types, and is associated with cell apoptosis, tumor invasion, metastasis, and chemoresistance in PDAC. In this study, we showed that both HIF-1α and CypA expression were significantly associated with lymph node metastasis and tumor stage. The expression of CypA was correlated with HIF-1α. Moreover, the mRNA and protein expression of CypA markedly decreased or increased following the suppression or over-expression of HIF-1α in vitro. Chromatin immunoprecipitation analysis showed that HIF-1α could directly bind to the hypoxia response element (HRE) in the CypA promoter regions and regulated CypA expression. Consistent with other studies, HIF-1α and CypA promoted PDAC cell proliferation and invasion, and suppressed apoptosis in vitro. Furthermore, we proved the combination effect of 2-methoxyestradiol and cyclosporin A both in vitro and in vivo. These results suggested that,CypA, a gene downstream of HIF-1α, could promote the development of PDAC. Thus, CypA might serve as a potential therapeutic target for PDAC.
Sigma-Aldrich
MISSION® esiRNA, targeting human PPIA
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
500 g
Please contact Customer Service for Availability
₩113,883
1 kg
In Stock
Details...
₩143,420