Merck
  • CMX001 (1-O-hexadecyloxypropyl-cidofovir) inhibits polyomavirus JC replication in human brain progenitor-derived astrocytes.
CMX001 (1-O-hexadecyloxypropyl-cidofovir) inhibits polyomavirus JC replication in human brain progenitor-derived astrocytes.
Antimicrobial agents and chemotherapy (2011-03-16)
Rainer Gosert, Christine Hanssen Rinaldo, Marion Wernli, Eugene O Major, Hans H Hirsch
ABSTRACT
Polyomavirus JC (JCV) replication causes progressive multifocal leukoencephalopathy (PML), a frequently fatal brain disease in immunodeficient patients, yet antiviral drugs are lacking. We characterized the lipid conjugate 1-O-hexadecyloxypropyl-cidofovir (CMX001) regarding JCV (Mad-4) replication in human brain progenitor-derived astrocytes (PDA) and the simian virus 40 (SV40) large-T-antigen-expressing COS-7 cells up to 7 days postinfection (dpi). We examined JCV loads by PCR, the infection rate by immunofluorescence, and host cell toxicity by WST-1 and BrdU incorporation assays. Supernatants from CMX001-treated PDA demonstrated a drug concentration-dependent decrease in JCV loads and infectivity. CMX001 had only a modest effect on host cell metabolism but reduced overall BrdU incorporation. In PDA at 7 dpi, the CMX001 50% effective concentration (EC50) was 5.55 nM, the 50% cytotoxic concentration (CC50) was 184.6 nM, and the 50% selectivity index (SI50) was 33.3. The EC90 was 19.7 nM, the CC90 was 5,054 nM, and the SI90 was 256.1. In COS-7 cells, JCV replication was faster and the EC50 and EC90 were 18- and 37-fold higher than those in PDA, i.e., 0.1 μM and 0.74 μM (CC50, 0.67 μM; SI50, 6.7; CC90, 12.2 μM; SI90, 16.5) at 5 dpi. We conclude that CMX001 inhibits JCV replication at concentrations in vitro that can be attained by oral administration without significant side effects in clinical studies.
Product Description
Roche
Cell Proliferation Reagent WST-1, suitable for protein quantification, suitable for cell analysis, detection, solution