Merck
  • Negative hemodynamic response without neuronal inhibition investigated by combining optical imaging and electrophysiological recording.
Negative hemodynamic response without neuronal inhibition investigated by combining optical imaging and electrophysiological recording.
Neuroscience letters (2016-11-20)
Zengguang Ma, Pengjia Cao, Pengcheng Sun, Zhuofan Lu, Liming Li, Yao Chen, Xinyu Chai
ABSTRACT
Understanding the mechanisms underlying negative hemodynamic responses is critical for the interpretation of functional brain imaging signals. Negative imaging signals have been found in the visual, somatosensory and motor cortices in functional magnetic resonance imaging (fMRI) and intrinsic signal optical imaging (ISOI) studies. However, the origin of negative imaging signals is still controversial. The present study investigated the visual cortical responses to peripheral grating stimuli using multi-wavelength ISOI and electrophysiological recording. We found an increased cerebral blood volume (CBV) in the stimulus-induced regions and a decreased CBV in the adjacent regions in the visual cortex. Nevertheless, there was no significant change in blood oxygenation in the negative CBV regions. Furthermore, by combining the planar and laminar electrophysiological recordings, we did not observe significantly decreased neuronal activity in the negative CBV regions. Our results suggest that the negative hemodynamic response does not necessarily originate in decreased neuronal activity. Therefore, caution should be taken when interpreting a negative hemodynamic response as neuronal inhibition.
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 50 cSt (25 °C)(lit.)