Merck
  • Caveolar remodeling is a critical mechanotransduction mechanism of the stretch-induced L-type Ca
Caveolar remodeling is a critical mechanotransduction mechanism of the stretch-induced L-type Ca
Pflugers Archiv : European journal of physiology (2017-03-18)
Sang Woong Park, Kyung Chul Shin, Hyun Ji Park, Soon-Kyu Yoou, Jin-Yeon Park, Young-Sun Kang, Dong Jun Sung, Jae Gon Kim, Seung Hwa Park, BoKyung Kim, Hana Cho, Young Min Bae
ABSTRACT
Activation of L-type voltage-dependent Ca
Sigma-Aldrich
MISSION® esiRNA, targeting human CAV1
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Estimated to ship on March 18, 2025
$192.00