Merck
  • Modulation of pentose phosphate pathway during cell cycle progression in human colon adenocarcinoma cell line HT29.
Modulation of pentose phosphate pathway during cell cycle progression in human colon adenocarcinoma cell line HT29.
International journal of cancer (2009-03-03)
Pedro Vizán, Gema Alcarraz-Vizán, Santiago Díaz-Moralli, Olga N Solovjeva, Wilma M Frederiks, Marta Cascante
ABSTRACT
Cell cycle regulation is dependent on multiple cellular and molecular events. Cell proliferation requires metabolic sources for the duplication of DNA and cell size. However, nucleotide reservoirs are not sufficient to support cell duplication and, therefore, biosynthetic pathways should be upregulated during cell cycle. Here, we reveal that glucose-6-phosphate dehydrogenase (G6PDH) and transketolase (TKT), the 2 key enzymes of oxidative and nonoxidative branches of the pentose phosphate pathway (PPP), respectively, which is necessary for nucleotide synthesis, are enhanced during cell cycle progression of the human colon cancer cell line HT29. These enhanced enzyme activities coincide with an increased ratio of pentose monophosphate to hexose monophosphate pool during late G1 and S phase, suggesting a potential role for pentose phosphates in proliferating signaling. Isotopomeric analysis distribution of nucleotide ribose synthesized from 1,2-(13)C(2)-glucose confirms the activation of the PPP during late G1 and S phase and reveals specific upregulation of the oxidative branch. Our data sustain the idea of a critical oxidative and nonoxidative balance in cancer cells, which is consistent with a late G1 metabolic check point. The distinctive modulation of these enzymes during cell cycle progression may represent a new strategy to inhibit proliferation in anticancer treatments.
Sigma-Aldrich
D-(−)-Ribose, suitable for cell culture, BioReagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
₩136,112
100 g
In Stock
Details...
₩374,028