Key Documents
Z742519
Block for digital dry bath and MyBlock
Holds 12 x 5.0 mL tubes
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41102404
NACRES:
NB.77
특징
Holds 12 x 5.0 mL tubes
포장
pack of 1 ea
제조업체/상표
Benchmark Scientific BSW5MT
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
법적 정보
MyBlock is a trademark of Benchmark Scientific, Inc.