콘텐츠로 건너뛰기
MilliporeSigma
모든 사진(2)
주요 문서
1.17340
Millipore
MAS-100 Atmos® Dust Cover
for use with MAS-100 Atmos®
로그인조직 및 계약 가격 보기
크기 선택
1173400001
$106.00
$106.00
예상 입고일April 21, 2025
추가 정보 요청
크기 선택
보기 변경
1173400001
$106.00
About This Item
UNSPSC 코드:
41103403
NACRES:
NA.74
$106.00
예상 입고일April 21, 2025
추가 정보 요청
material
white polypropylene cover
설명
Compressed gas sampling
특징
autoclavable
제조업체/상표
MAS-100 Atmos®
응용 분야
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
호환성
for use with MAS-100 Atmos®
유사한 품목 비교
전체 비교 보기
차이점 표시
1 of 4
이 품목
1.173361.090841.17349
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
application(s)
air monitoring, cosmetics
food and beverages
pharmaceutical
application(s)
compressed gas monitoring
cosmetics
food and beverages
pharmaceutical
material
white polypropylene cover
material
aluminum cover
material
aluminum cover
material
PVC tubing, stainless steel adapter
description
Compressed gas sampling
description
Compressed gas sampling
description
-
description
-
feature
autoclavable
feature
autoclavable
feature
autoclavable
feature
autoclavable
manufacturer/tradename
MAS-100 Atmos®
manufacturer/tradename
MAS-100 Atmos®
manufacturer/tradename
-
manufacturer/tradename
MAS-100 Atmos®
일반 설명
MAS-100 Atmos® Dust Cover is an accessory to protect the instrument from dust. If the perforated lid is dismantled for autoclaving, it is recommended to protect the device with the MAS-100 Atmos® dust cover.
The dust cover is white and made from polypropylene. It can be sterilized by steam sterilization (autoclaving) at 134°C for 5 minutes or 121°C for 20 minutes. Disinfection is done by spraying or wiping with 70% Ethanol or 70% IPA or other disinfectants compatible with polypropylene.
애플리케이션
The MAS-100 Atmos® Dust Cover is used to prevent dust accumulation on the sampling head of MAS-100 Atmos® Microbial Compressed Gas Sampler.
분석 메모
The MAS-100 Atmos® dust cover will slightly shrink after the first autoclaving cycle, resulting in some clearance. This should be considered during transport but has no effect on the protective capacity of the cover.
법적 정보
MAS-100 Atmos is a registered trademark of MBV AG