Key Documents
Z290750
Sigma-Aldrich
Vacuum filtration assembly funnel stems
no membrane support, membrane diam. 47 mm, connector for stopper
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104900
NACRES:
NB.24
material
glass housing
특징
no membrane support
포장
pkg of 1 ea
멤브레인 직경
47 mm
피팅
connector for stopper