콘텐츠로 건너뛰기
Merck
  • Crossing the barrier: net flux of 27-hydroxycholesterol into the human brain.
Crossing the barrier: net flux of 27-hydroxycholesterol into the human brain.
Journal of lipid research (2005-03-03)
Maura Heverin, Steve Meaney, Dieter Lütjohann, Ulf Diczfalusy, John Wahren, Ingemar Björkhem
초록
Side chain oxidized oxysterols have a unique ability to traverse lipophilic membranes. We tested the hypothesis that there is a net flux of 27-hydroxycholesterol from the circulation into the brain using plasma samples collected from the internal jugular vein and an artery of healthy male volunteers. Two independent studies were performed, one in which total levels of 27-hydroxycholesterol were measured and one in which the free fraction of 27-hydroxycholesterol was measured. In the majority of subjects studied, the level of 27-hydroxycholesterol was higher in the artery than in the vein, and uptake from the circulation was calculated to be about 5 mg/24 h. The distribution of 27-hydroxycholesterol in human brain was found to be consistent with an extracerebral origin, with a concentration gradient from the white to the gray matter--a situation opposite that of 24S-hydroxycholesterol, which os exclusively formed in brain. In view of the fact that the blood-brain barrier is impermeable to cholesterol and that 27-hydroxycholesterol is a potent regulator of several cholesterol-sensitive genes, the flux of 27-hydroxycholesterol into the brain may be and important link between intra- and extracerebral cholesterol homeostasis.
MATERIALS
제품 번호
브랜드
제품 설명
Avanti
27-hydroxycholesterol, Avanti Research - A Croda Brand