Syringe Filters for Chromatography
We carry both Millex® and Whatman® syringe filters, the two premier brands in syringe filters for chromatography. Our combined portfolio offers many different membranes and housings for use in a wide range of applications.
Syringe Filter Selection Guide for Chromatography
The presence of particles in chromatography samples can cause pressure build up in the system, leading to instrument shutdown. This is especially true in UHPLC, due to its small particle-packed columns and narrow bore tubing. Regardless of the application, this potential problem is easily eliminated by sample filtration.
Application | HPLC | UPLC®/UHPLC | Ion Chromatography/IC |
---|---|---|---|
Recommended Pore Size | 0.45 μm syringe filter | 0.2 μm syringe filter | 0.45 μm or 0.2 μm syringe filters |
Recommend Diameter by Sample Volume | <1 mL: 4 mm 1-10 mL: 13 mm 10-100 mL: 30-33 | <1 mL: 4 mm 1-10 mL: 13 mm 10-100 mL: 30-33 mm | 1-10 mL: 13 mm 10-100 mL: 25 mm |
Chemical Compatibility | Broad chemical compatibility (aqueous and organic) | Broad chemical compatibility (aqueous and organic) | Broad chemical compatibility (aqueous) |
Extractables | Low extractables prevent sample contamination | Low extractables prevent sample contamination | Low levels of ionic extractables prevent sample contamination |
View recommended syringe filters for HPLC | View recommended syringe filters for UHPLC | View recommended syringe filters for IC |
Recommended Products by Application
Syringe Filters for HPLC
Sample volume | Membrane Type | Product Description | Product Number |
---|---|---|---|
<1 mL | PTFE, hydrophilic | Millex®-LCR 0.45µm hydrophilic PTFE syringe filter, 4mm | SLLHR04NL |
1-10 mL | PTFE, hydrophilic | Millex®-LCR 0.45µm hydrophilic PTFE syringe filter, 13mm | SLCRX13 |
Regenerated Cellulose | Whatman Spartan® HPLC certified syringe filters, 13mm | WHA10463110 | |
10-100 mL | |||
PTFE, hydrophilic | Millex®-LCR 0.45µm hydrophilic PTFE syringe filter, 33mm | SLCR033 | |
Regenerated Cellulose | Whatman Spartan® HPLC certified syringe filters, 30mm | WHA10463053 |
Syringe Filters for UPLC® / UHPLC
Sample volume | Membrane Type | Product Description | Product Number |
---|---|---|---|
<1 mL | PTFE, hydrophilic | Millex®-LG 0.20µm hydrophilic PTFE syringe filter, 4mm | SLLGR04NL |
1-10 mL | PTFE, hydrophilic | Millex®-LG 0.20µm hydrophilic PTFE syringe filter, 13mm | SLLGX13 |
Regenerated Cellulose | Whatman Spartan® HPLC certified syringe filters, 13mm | WHA10463100 | |
10-100 mL | |||
PTFE, hydrophilic | Millex®-LCR 0.20µm hydrophilic PTFE syringe filter, 33mm | SLLG033 | |
Regenerated Cellulose | Whatman Spartan® HPLC certified syringe filters, 30mm | WHA10463060 |
Syringe Filters for Ion Chromatography/IC
Sample volume | Membrane Type | Product Description | Product Number |
---|---|---|---|
1-10 mL | PTFE, hydrophilic | Millex®-LH 0.45µm hydrophilic PTFE syringe filter, 13mm | SLLHC13NL |
PTFE, hydrophilic | Millex®-LH 0.20µm hydrophilic PTFE syringe filter, 13mm | SLLGC13NL | |
Anopore® | Whatman Anotop® IC and LC syringe filter, Anotop® 10 IC Syringe Filter, pore size 0.2 μm, polypropylene housing, | WHA68099233 | |
10-100 mL | PTFE, hydrophilic | Millex®-LH 0.45µm hydrophilic PTFE syringe filter, 25mm | SLLHC25NS |
PTFE, hydrophilic | Millex®-LH 0.20µm hydrophilic PTFE syringe filter, 25mm | SLLGC25NS | |
Anopore® | Whatman Anotop® IC and LC syringe filter, Anotop® 25 LC Syringe Filter, pore size 0.2 μm, polypropylene housing, | WHA20025100 |