MilliporeSigma
Sterility Test Sampling Solutions
Steritest® needle guide and Steritest® needle-free valve
Request a Free Sample or Get More Information About Our Sterility Test Sampling Solutions
While performing the compendial sterility test, you may need to access the sample in the canister during incubation. For example, you may need to subculture, reincubate or identify microorganisms that may have grown in turbid culture medium, or to inject a liquid.
You have 2 solutions to access the sample: the needle guide for unplanned sampling on short notice or the needle-free valve for scheduled sampling or just in case.