Iupac Name
zinc diacrylate
Inchi Code
1S/2C3H4O2.Zn/c2*1-2-3(4)5;/h2*2H,1H2,(H,4,5);/q;;+2/p-2
InChI key
XKMZOFXGLBYJLS-UHFFFAOYSA-L
storage temp.
RT
Purity
98%
Country of Origin
US
Product Link
https://www.aablocks.com/prod/14643-87-9
MSDS
https://www.aablocks.com/downloadSds?cas=14643-87-9
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service