Skip to Content
MilliporeSigma

AABH97D211A9

AA BLOCKS, INC.

naphthalene-1-sulfonic acid hydrate

Synonym(s):

naphthalene-1-sulfonic acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O4S1
CAS Number:
Molecular Weight:
226.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H8O3S.H2O/c11-14(12,13)10-7-3-5-8-4-1-2-6-9(8)10;/h1-7H,(H,11,12,13);1H2

Inchi Key

DCNHVBSAFCNMBK-UHFFFAOYSA-N

Storage temp.

RT

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library