Skip to Content
MilliporeSigma

AABH99A50E21

AA BLOCKS, INC.

Boc-NH-PEG4-C2-Boc

Synonym(s):

tert-butyl 2,2-dimethyl-4-oxo-3,8,11,14,17-pentaoxa-5-azaicosan-20-oate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H39O8N1
CAS Number:
Molecular Weight:
421.53
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H39NO8/c1-19(2,3)28-17(22)7-9-24-11-13-26-15-16-27-14-12-25-10-8-21-18(23)29-20(4,5)6/h7-16H2,1-6H3,(H,21,23)

Inchi Key

HOBFRUPMGLPRSJ-UHFFFAOYSA-N

Purity

98%

Storage temp.

2-8C

Country of Origin

US



Still not finding the right product?

Explore all of our products under Boc-NH-PEG4-C2-Boc




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library