Skip to Content
MilliporeSigma

AABH9A2286A1

AA BLOCKS, INC.

Trimethyl chitosan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H27O4N2Cl1
CAS Number:
Molecular Weight:
286.8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H24NO4.ClH.H3N/c1-7-8(6-13)16-11(15-5)9(10(7)14)12(2,3)4;;/h7-11,13-14H,6H2,1-5H3;1H;1H3/q+1;;/p-1/t7-,8-,9-,10+,11-;;/m1../s1

Inchi Key

HFNQAFWMDXYZFM-YHSFBLOESA-M

Purity

98%

Storage temp.

-20C

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library