Skip to Content
MilliporeSigma

AABH9A954262

AA BLOCKS, INC.

Leuprorelin (acetate)

Synonym(s):

(S)-N-ethyl-1-(((S)-5-oxopyrrolidine-2-carbonyl)-L-histidyl-L-tryptophyl-L-seryl-L-tyrosyl-D-leucyl-L-leucyl-L-arginyl)pyrrolidine-2-carboxamide acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C59H84N16O12.C2H4O2/c1-6-63-57(86)48-14-10-22-75(48)58(87)41(13-9-21-64-59(60)61)68-51(80)42(23-32(2)3)69-52(81)43(24-33(4)5)70-53(82)44(25-34-15-17-37(77)18-16-34)71-56(85)47(30-76)74-54(83)45(26-35-28-65-39-12-8-7-11-38(35)39)72-55(84)46(27-36-29-62-31-66-36)73-50(79)40-19-20-49(78)67-40;1-2(3)4/h7-8,11-12,15-18,28-29,31-33,40-48,65,76-77H,6,9-10,13-14,19-27,30H2,1-5H3,(H,62,66)(H,63,86)(H,67,78)(H,68,80)(H,69,81)(H,70,82)(H,71,85)(H,72,84)(H,73,79)(H,74,83)(H4,60,61,64);1H3,(H,3,4)/t40-,41-,42-,43+,44-,45-,46-,47-,48-;/m0./s1

Inchi Key

RGLRXNKKBLIBQS-XNHQSDQCSA-N

Purity

98%

Country of Origin

US

Storage temp.

N/A



Still not finding the right product?

Explore all of our products under Leuprorelin (acetate)




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library