Inchi Code
1S/C19H21BrFNO.BrH/c1-22(2)11-3-10-19(15-4-7-17(21)8-5-15)18-9-6-16(20)12-14(18)13-23-19;/h4-9,12H,3,10-11,13H2,1-2H3;1H
Inchi Key
WZLFJKIEXDRHDD-UHFFFAOYSA-N
Storage temp.
N/A
Country of Origin
US
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.