MilliporeSigma
AATB-17207
AAT Bioquest
AAT Bioquest ddCTP
1 umole 10 mM in ddH2O
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
Synonym(s):
2', 3'-Dideoxycytidine-5'-triphosphate
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
AAT Bioquest
General description
Sanger sequencing with 99.99% accuracy is still the “gold standard” for clinical research sequencing. dd-CTP is one of the four critical ddNTP components for performing Sanger sequencing.
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH2O* is manufactured by AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)