MilliporeSigma
AATB-20011
AAT Bioquest
AAT Bioquest HHBS
100 mL
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
Synonym(s):
Hanks' Buffer with 20 mM Hepes
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
AAT Bioquest
General description
Hank's Balanced Salt Solution with 20 mM Hepes buffer (HHBS) is used for a variety of cell culture applications, such as washing cells, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
HHBS [Hanks' Buffer with 20 mM Hepes] is manufactured by AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)