MilliporeSigma
AATB-22225
AAT Bioquest
AAT Bioquest DASPEI
100 mg
Shipped & Sold By AAT Bioquest
A Trusted Marketplace Seller
Synonym(s):
2-(4-(dimethylamino)styryl)-N-ethylpyridinium iodide
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
AAT Bioquest
General description
DASPEI is a styryl dye that stains mitochondria of live cells.
DASPEI [2-(4-(dimethylamino)styryl)-N-ethylpyridinium iodide] *CAS#: 3785-01-1* is manufactured by AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)